missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Smart-Vue™ Receivers

Choose from an assortment of Thermo Scientific™ Smart-Vue™ receivers including receiver packages that include power supply and software for your wireless monitoring solution.

Brand:  Thermo Scientific™ SV103-513-LSB

Additional Details : Poids : 0.37000kg

 View more versions of this product

Product Code. 15380321

  • 3582.00€ / Each


Expédition estimée
16-11-2020
Add to basket

Description

Description

Compatible with:

Refrigerators, Freezers, LN2 Storage, CO2 Incubators, Clean Rooms, Storage Areas, Ovens

Ordering Information:

Availability of Smart-Vue products vary by country. Please ask your local Sales Representative to evaluate a Smart-Vue wireless monitoring solution that best meets your specific requirements. Specifications, terms and pricing are subject to change. Not all products are available in all countries.

Specifications

Specifications

Smart-Vue facilitates conformance to 21 CFR Part 11, cGxP and SOP compliance.
Smart-Vue™ Wireless Monitoring Solutions
868MHz
USB 1.0 or 2.0 Receiver, 25mW+SW 99 sensors
Velcro, CD ROM with Smart-Vue client server software 2.0 license up to 99 sensors, software drivers and user manuals.
Receiver, 868MHz
Videos
Documents