Learn More
Sartorius Smooth Technical Filter Papers
This Smooth Technical Filter Paper grade is generally used for analytical and technical purposes.
13.00€ - 308.00€
Specifications
Material Cotton Linters, Refined Pulp
Material (Membrane) Cellulose
Type Smooth Filter Paper
Fit for Purpose Certified Quality
View More Specs
Products 4
Product Code Brand Dimensions (L x W) Grade Price Quantity & Availability  
Product Code Brand Dimensions (L x W) Grade Price Quantity & Availability  
15952720
View Documents Promotion Details
Sartorius
FT-1-H079-040020
40 x 20 mm (1.6 x 0.8 in.) H079
65.40€
Each
View alternative products
Save Up to 
»
Estimated Shipment: 13-05-2024
to see stock.
Add to basket
15902780
View Documents Promotion Details
Sartorius
FT-2-356-089140
89 × 140 mm (3.5 x 5.5 in.) C 140
13.00€
Each
View alternative products
Save Up to 
»
Estimated Shipment: 13-05-2024
to see stock.
Add to basket
15922780
View Documents Promotion Details
Sartorius
FT-2-356-445572
445 × 572 mm (17.5 x 22.5 in.) C 140
308.00€
Each
View alternative products
Save Up to 
»
Estimated Shipment: 13-05-2024
to see stock.
Add to basket
15952760
View Documents Promotion Details
Sartorius
FT-2-326-160210
160 x 210 mm (6.3 x 8.3 in.) 69 K This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
N/A View alternative products
Save Up to 
»
Description
Description
  • Made of refined pulp and cotton linters with a >95% α-cellulose content
  • Ash content between 0.1 to 0.15%
  • Wet-strengthened
Specifications
Specifications
Cotton Linters, Refined Pulp
Cellulose
Smooth Filter Paper
Certified Quality