Learn More
Macherey-Nagel™ Snap Tubes
Click to view available options
Volume (Metric) Well:
15 mL
50 mL
Unit Size:
Pack of 50
Specifications
Specifications
| Color | Clear |
| Format | Conical Tube |
| For Use With (Equipment) | Centrifuge |
| For Use With (Application) | Incubation and lysis of samples |
| Includes | Snap lid |
| Product Type | Centrifuge Tube |
| Quantity | 50 Pack |
| Volume (Metric) Well | 50 mL |