missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SNaPshot™ Multiplex Kit
Shop All Applied Biosystems Products
Click to view available options
No. of Reactions:
1000
5000
Unit Size:
Each
Description
Use this system to screen and confirm SNPs, detect minor sequence variations, fingerprint bacterial artificial chromosomes.
Order Info
Each
Specifications
Specifications
| Accession Number | Capillary, 61cm Capillary Length, Sequencing, Room Temperature, For 310 Genetic Analyzer |
| Description | SNaPshot™ Multiplex Kit |
| No. of Reactions | 1000 |
For research use only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction