Learn More
Brannan™ Specific Gravity Hydrometers M100 Series
Click to view available options
Specific Gravity Range:
0.70 to 0.80 g/mL
0.80 to 0.90 g/mL
0.90 to 1.00 g/mL
1.10 to 1.20 g/mL
1.20 to 1.30 g/mL
1.30 to 1.40 g/mL
Unit Size:
Each
Description
Compares the relative density of a given liquid with that of water at 15.6°C (60°F). Brannan™ M100 Series Specific Gravity Hydrometers are manufactured from soda glass, steel shot ballast and synthetic wax resin.
- Includes protective plastic tube
- 250 mm in length
- Scale subdivision 0.002
- Accuracy of 0.002 specific gravity
- Available adjusted for either low or medium surface tension
- Available for numerous specific gravity ranges
Specifications
Specifications
| Quantity | 1 Ea. |
| Type | Specific Gravity Hydrometer |
| Accuracy | 0.002 g/mL |
| Increments | 0.002 g/mL |
| Certifications/Compliance | ISO 650, 916 |
| Length (Metric) | 250 mm |
| Ballast Type | Steel Shot |
| For Use With (Application) | Relative density of a given liquid with that of water |
| Specific Gravity Range | 1.20 to 1.30 g/mL |