Learn More
Rogo Sampaic™ Anumbra™ Glass Petri Dishes
Product Code. p-8009977
Click to view available options
Diameter (Metric):
60 mm
90 mm
100 mm
120 mm
150 mm
200 mm
Height (Metric):
12 mm
15 mm
20 mm
25 mm
Unit Size:
Pack of 10
Pack of 20
Product Code. 11733923
Brand: Rogo Sampaic™ BRB002
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Excellent for microbial or cell culture applications in the laboratory. Rogo Sampaic™ Anumbra™ Glass Petri Dishes are manufactured from surface-hardened soda lime glass.
  • Reusable
  • Autoclavable
  • Available in a range of diameters and heights
Specifications
Material Soda Lime Glass
Shape Round
Type Petri Dish
Height (Metric) 12 mm
Diameter (Metric) 60 mm