Learn More
Hirschmann™ Soda-lime Glass Test Tube with Round Bottoms
Click to view available options
Diameter (Metric):
12 mm
16 mm
Length (Metric):
75 mm
100 mm
125 mm
150 mm
160 mm
180 mm
missing translation for 'unitSize'
Pack of 100
Description
- Plain end
- Round bottom
- Sodalime-glass
Specifications
Specifications
| Length (Metric) | 100 mm |
| Material | Soda Lime Glass |
| Packaging | 1 Pack |
| Quantity | 1 Pack |
| Diameter (Metric) | 12 mm |
| Product Type | Test Tube |