missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Acetate, ACS Reagent, Honeywell Fluka™
Click to view available options
Quantity:
5 g
100 g
500 g
1 kg
2.5 kg
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Quantity | 5 g |
Linear Formula | CH3COONa |
UN Number | NONH for all modes of transport |
Beilstein | 3595639 |
Flash Point | >250°C (>482°F) |
Formula Weight | 82.03g/mol |
Percent Purity | ≥99% |
Grade | ACS Reagent |
Ignition Point | 1112°F |
Chemical Name or Material | Sodium Acetate |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'