missing translation for 'onlineSavingsMsg'
Learn More

Sodium Azide, ≥99%

Product Code. 15835188 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
Packaging:
Plastic bottle
Unit Size:
100g
1kg
500g
Product Code. Quantity Packaging unitSize
15835188 100 g Plastic bottle 100g
15845188 500 g Plastic bottle 500g
15895328 1 kg Plastic bottle 1kg
3 options
This item is not returnable. View return policy

Product Code. 15835188

Brand: Thermo Scientific J21610.22

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 26628-22-8 | N3Na | 65.011 g/mol

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 26628-22-8
Molecular Formula N3Na
Molecular Weight (g/mol) 65.011
MDL Number MFCD00003536
InChI Key PXIPVTKHYLBLMZ-UHFFFAOYSA-N
Synonym sodium azide, azide, sodium, natriumazid, nemazyd, smite, kazoe, azoture de sodium, sodium azide na n3, hydrazoic acid, sodium salt, natriumazid german
PubChem CID 33557
ChEBI CHEBI:278547
IUPAC Name sodium;azide
SMILES [N-]=[N+]=[N-].[Na+]

Specifications

Melting Point ∼275°C (decomposition)
Physical Form Crystalline Powder
Quantity 100 g
UN Number UN1687
Formula Weight 65.01
Percent Purity ≥99%
Packaging Plastic bottle
Specific Gravity 1.846
Density 1.846
Chemical Name or Material Sodium Azide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.