Learn More
Thermo Scientific Chemicals Sodium benzoate, 99%, for biochemistry
Catalog Number p-4813641
Click to view available options
Quantity:
250 g
1 kg
532-32-1
C7H5NaO2
144.11
MFCD00012463
WXMKPNITSTVMEF-UHFFFAOYSA-M
sodium benzoate, benzoic acid, sodium salt, benzoic acid sodium salt, sobenate, antimol, benzoate sodium, benzoate of soda, benzoate, sodium, natrium benzoicum, caswell no. 746
517055
sodium;benzoate
[Na+].[O-]C(=O)C1=CC=CC=C1
532-32-1
C7H5NaO2
144.11
MFCD00012463
WXMKPNITSTVMEF-UHFFFAOYSA-M
sodium benzoate, benzoic acid, sodium salt, benzoic acid sodium salt, sobenate, antimol, benzoate sodium, benzoate of soda, benzoate, sodium, natrium benzoicum, caswell no. 746
517055
sodium;benzoate
[Na+].[O-]C(=O)C1=CC=CC=C1
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Melting Point >300.0°C
Assay Percent Range 99%
Packaging Plastic bottle
Linear Formula C6H5CO2Na
Quantity 250 g
Merck Index 15,8718
Solubility Information (1 M in water) Clear colorless to light yellow
Formula Weight 144.11
Percent Purity 99%
Grade Biochemistry
Chemical Name or Material Sodium benzoate
Show More Show Less
compliance-icons
Product Identifier
  • Benzoic acid, sodium salt
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.