Learn More
Honeywell Fluka™ Sodium bisulfate, Honeywell Fluka™
Purum, anhydrous, ~95.0%
Brand: Honeywell Fluka™ 71656-1KG
Additional Details : CAS : 7681-38-1 Poids : 1.00000kg
Quantity | 1kg |
---|
Description
- ≥95% for general laboratory use.
Chemical Identifiers
7681-38-1 | |
120.054 | |
WBHQBSYUUJJSRZ-UHFFFAOYSA-M | |
516919 | |
OS(=O)(=O)[O-].[Na+] |
HNaO4S | |
MFCD00003531 | |
sodium hydrogen sulfate, sodium bisulfate, sodium acid sulfate, bisulfate of soda, sodium hydrosulfate, fanal, sodium hydrogensulphate, niter cake, nitre cake, wc-perfect | |
sodium;hydrogen sulfate |
Specifications
Sodium bisulfate | |
HNaO4S | |
MFCD00003531 | |
sodium hydrogen sulfate, sodium bisulfate, sodium acid sulfate, bisulfate of soda, sodium hydrosulfate, fanal, sodium hydrogensulphate, niter cake, nitre cake, wc-perfect | |
OS(=O)(=O)[O-].[Na+] | |
120.054 | |
120.06g/mol | |
1kg |
7681-38-1 | |
NaHSO4 | |
NONH for all modes of transport | |
WBHQBSYUUJJSRZ-UHFFFAOYSA-M | |
sodium;hydrogen sulfate | |
516919 | |
93 to 102% (T), 95% |