missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium bisulfate, Honeywell Fluka™
Click to view available options
Quantity:
250 g
1 kg
5 kg
Description
- ≥95% for general laboratory use.
Specifications
Quantity | 250 g |
Linear Formula | NaHSO4 |
UN Number | NONH for all modes of transport |
Formula Weight | 120.06g/mol |
Percent Purity | 93 to 102% (T), 95% |
Chemical Name or Material | Sodium bisulfate |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'