missing translation for 'onlineSavingsMsg'
Learn More

Sodium bromide, ACS reagent, Thermo Scientific Chemicals

Catalog Number p-4380031
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Plastic bottle
This item is not returnable. View return policy
7647-15-6
BrNa
102.89
MFCD00003475
JHJLBTNAGRQEKS-UHFFFAOYSA-M
sodium bromide, bromide salt of sodium, sodium bromide nabr, sedoneural, sodiumbromide, trisodium tribromide, bromnatrium, nabr, bromnatrium german, caswell no. 750a
253881
CHEBI:63004
[Na+].[Br-]
This item is not returnable. View return policy
7647-15-6
BrNa
102.89
MFCD00003475
JHJLBTNAGRQEKS-UHFFFAOYSA-M
sodium bromide, bromide salt of sodium, sodium bromide nabr, sedoneural, sodiumbromide, trisodium tribromide, bromnatrium, nabr, bromnatrium german, caswell no. 750a
253881
CHEBI:63004
[Na+].[Br-]

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Boiling Point 1390.0°C
Melting Point 755.0°C
Quantity 100 g
Linear Formula NaBr
Fieser 12,445; 13,46
Merck Index 15,8729
Solubility Information Solubility in water: 905g/L (20°C). Other solubilities: 1160g/L water (50°C), 1210g/L water (100°C), 1g/6 mL methanol, 7g/100 mL 95% ethanol
IUPAC Name sodium bromide
Formula Weight 102.89
Grade ACS Reagent
Packaging Glass bottle
Chemical Name or Material Sodium bromide
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'