missing translation for 'onlineSavingsMsg'
Learn More

Sodium Carbonate, ACS Reagent (primary standard), Honeywell™

Product Code. 15602220 Shop All Honeywell Fluka Products
Click to view available options
Quantity:
500 g
Unit Size:
500g
This item is not returnable. View return policy

Product Code. 15602220

Brand: Honeywell-Fluka 223484500G

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 497-19-8
Physical Form Anhydrous
Quantity 500 g
Molecular Formula CNa2O3
Linear Formula Na2CO3
MDL Number MFCD00003494
UN Number NONH for all modes of transport
Beilstein 4154566
Synonym sodium carbonate, disodium carbonate, soda ash, carbonic acid disodium salt, calcined soda, sodium carbonate, anhydrous, carbonic acid, disodium salt, washing soda, sodiumcarbonate, solvay soda
InChI Key CDBYLPFSWZWCQE-UHFFFAOYSA-L
SMILES [Na+].[Na+].[O-]C([O-])=O
IUPAC Name disodium carbonate
Molecular Weight (g/mol) 105.99
PubChem CID 10340
ChEBI CHEBI:29377
Formula Weight 105.99g/mol
Percent Purity 99.95 to 1005% (dry basis)
Grade ACS Reagent
Chemical Name or Material Sodium Carbonate
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.