missing translation for 'onlineSavingsMsg'
Learn More

Sodium fluoride, Puriss. p.a., ACS Reagent, Reag. ISO, Reag. Ph. Eur., ≥99%, Honeywell Fluka™

Catalog Number p-7071960
Click to view available options
Quantity:
250 g
500 g
1 kg
6 x 250 g
50 kg
6 x 1 kg
This item is not returnable. View return policy
7681-49-4
FNa
41.99
MFCD00003524
PUZPDOWCWNUUKD-UHFFFAOYSA-M
sodium fluoride, florocid, fluoride, sodium, zymafluor, ossin, sodium fluoride naf, sodium fluoride, antibulit, floridine, fluonatril
5235
CHEBI:28741
[F-].[Na+]
This item is not returnable. View return policy
7681-49-4
FNa
41.99
MFCD00003524
PUZPDOWCWNUUKD-UHFFFAOYSA-M
sodium fluoride, florocid, fluoride, sodium, zymafluor, ossin, sodium fluoride naf, sodium fluoride, antibulit, floridine, fluonatril
5235
CHEBI:28741
[F-].[Na+]

Puriss. p.a., ACS Reagent, Reag. ISO, Reag. Ph. Eur.,≥99%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY
Quantity 250 g
Linear Formula NaF
UN Number 1690
IUPAC Name sodium fluoride
Formula Weight 41.99g/mol
Percent Purity ≥99%
Grade ACS/puriss. p.a.
Vapor Pressure 1.4mmHg (0°C)
Chemical Name or Material Sodium Fluoride, Reag. Ph. Eur. reag. ISO
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'