missing translation for 'onlineSavingsMsg'
Learn More

Sodium Hydroxide Solution, 0.25M, Chem-Lab

Product Code. 11953223
Click to view available options
Quantity:
1 L
Unit Size:
1 L
This item is not returnable. View return policy

Product Code. 11953223

Brand: Chem-Lab CL05.1414.1000

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Used in laboratory applications and in titrations. Sodium Hydroxide Solution, 0.25M, Chem-Lab is a strong base and is highly caustic. It is used to increase the alkalinity of a mixture, to neutralize acids and to make sodium salts. One liter of the solution includes 10g of NaOH.

Sodium hydroxide is used in many industries, mostly as a strong chemical base in the manufacture of pulp and paper, textiles, drinking water, soaps and detergents and as a drain cleaner.

Specifications

pH >12
Color Colorless
Boiling Point 100°C
CAS 1310-73-2, 7732-18-5
Molecular Formula HNaO
Concentration 0.25M
Density 1.01g/mL
Synonym sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
Solubility Information Soluble in Water
InChI Key HEMHJVSKTPXQMS-UHFFFAOYSA-M
Quantity 1 L
SMILES [OH-].[Na+]
IUPAC Name sodium;hydroxide
Molecular Weight (g/mol) 39.997
PubChem CID 14798
ChEBI CHEBI:32145
Formula Weight 40.00g/mol
Concentration or Composition (by Analyte or Components) 10g NaOH per 1L H2O
Normality 0.25 N (±0.0005, 20°C)
Odor Odorless
Solvent or Matrix Water
Physical Form Liquid
Show More Show Less
compliance-icons
Product Identifier
  • sodium hydroxide 0.25 mol/l
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H290-May be corrosive to metals.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P311-IF exposed or concerned: Call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : sodium hydroxide
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.