Learn More
Sodium hydroxide, 1.0N Standardized Solution
Description
Sodium hydroxide, 1.0N standardized solution is used for volumetric analysis in laboratories.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Color | Colorless |
| Physical Form | Liquid |
| Quantity | 20 L |
| Solubility Information | Miscible with glycerol,ethanol and methanol. Immiscible with acetone and ether. |
| Formula Weight | 40 |
| Appearance | Colourless liquid |
| Vapor Pressure | 23 hPa (17mm Hg) at 20°C |
| Chemical Name or Material | Sodium hydroxide |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.