Learn More
Honeywell Fluka™ Sodium hydroxide concentrate, For 1 L standard solution, 0.5 M NaOH (0.5 N), Honeywell Fluka™
for 1 L standard solution, 0.5 M NaOH (0.5 N)
Brand: Honeywell Fluka™ 38217-1EA
Additional Details : CAS : 1310-73-2 Poids : 1.43000kg Transport : UN number : 1824 Chem class : 8 Pack group : II
Quantity | 1 L |
---|
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Chemical Identifiers
1310-73-2 | |
39.997 | |
HEMHJVSKTPXQMS-UHFFFAOYSA-M | |
14798 | |
sodium;hydroxide |
HNaO | |
MFCD00003548 | |
sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic | |
CHEBI:32145 | |
[OH-].[Na+] |
Specifications
Sodium hydroxide concentrate | |
HNaO | |
MFCD00003548 | |
sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic | |
[OH-].[Na+] | |
39.997 | |
CHEBI:32145 | |
Ampoule | |
1 L |
1310-73-2 | |
NaOH | |
UN1824 | |
HEMHJVSKTPXQMS-UHFFFAOYSA-M | |
sodium;hydroxide | |
14798 | |
40g/mol | |
3 mmHg (37°C) |