missing translation for 'onlineSavingsMsg'
Learn More

Sodium Hydroxide Solution, ≥20%, Honeywell Fluka™

Catalog Number p-7071971
Click to view available options
Quantity:
1 L
2.5 L
25 L
This item is not returnable. View return policy
1310-73-2
HNaO
39.997
MFCD00003548
HEMHJVSKTPXQMS-UHFFFAOYSA-M
sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
14798
CHEBI:32145
sodium;hydroxide
[OH-].[Na+]
This item is not returnable. View return policy
1310-73-2
HNaO
39.997
MFCD00003548
HEMHJVSKTPXQMS-UHFFFAOYSA-M
sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
14798
CHEBI:32145
sodium;hydroxide
[OH-].[Na+]

≥20%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY
Linear Formula NaOH
Concentration ≥20%
UN Number 1824
Quantity 1 L
Formula Weight 40g/mol
Vapor Pressure 3mmHg (37°C)
Chemical Name or Material Sodium Hydroxide Solution
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'