missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Hydroxide Solution, Volumetric, 4M NaOH (4N), Honeywell™ Fluka™
Shop All Honeywell Fluka Products
Click to view available options
Quantity:
1 L
Packaging:
Poly bottle
Unit Size:
1 L
Description
Honeywell´s high-quality titration products cover the complete titration workflow and include:
- Solutions for volumetric titration (acids, bases, or salts)
- Buffers available as concentrates or ready-to-use
- Reagents for complexometry:
- Aminopolycarboxylic acids (EDTA/NTA analogs)
- Masking agents
- Indicators for volumetric and complexometric titrations
Specifications
Specifications
| pH | Alkaline |
| Color | Colorless |
| Boiling Point | 105°C to 140°C |
| CAS | 1310-73-2 |
| Melting Point | -10°C to +14°C |
| Molecular Formula | HNaO |
| Linear Formula | NaOH |
| MDL Number | MFCD00003548 |
| UN Number | UN1824 |
| Density | ∽1.15g/cu.cm at 20°C |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction