Learn More
Sodium Hypochlorite Solution, 12/13%, Chem Lab™
Product Code. 11921397
Click to view available options
Quantity:
1 L
Unit Size:
1 L
Product Code. 11921397
Brand: Chem-Lab CL02.1438.1000
Specifications
Color Yellow
Boiling Point 100°C
Melting Point 0°C
Composition NaClO/H2O (± 3% Act. Cl)
Density 1.04g/mL
Solubility Information Water Soluble
Odor Chlorine
pH >13
Quantity 1 L
Physical Form Liquid
compliance-icons
Product Identifier
  • Sodium hypochlorite 12/13% solution a.r.
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • EUH031-Contact with acids liberates toxic gas.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : sodium hypochlorite