missing translation for 'onlineSavingsMsg'
Learn More

Sodium hypophosphite hydrate, Honeywell Fluka™

Catalog Number 15658340
missing translation for 'orderingAttributeHoverText'
Quantity:
500 g
Packaging:
Poly Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy
CAS 123333-67-5
CAS Max % 100.0000%
Quantity 500 g
Molecular Formula H2NaO3P
Linear Formula CHCl3
MDL Number MFCD00000826
Synonym sodium hypophosphite hydrate, sodium phosphinate hydrate, sodium phosphenite hydrate, sodium hypophosphite en grade, sodium hydrate hypophosphite, sodium hypophosphite monohydrate 250g, phosphinic acid sodium salt monohydrate, sodium hypophosphite monohydrate, phosphonous acid monosodium salt monohydrate, sodium hypophosphite monohydrate, puriss
InChI Key ACUGTEHQOFWBES-UHFFFAOYSA-M
SMILES O.[O-]P=O.[Na+]
Molecular Weight (g/mol) 103.977
PubChem CID 23708894
Formula Weight 87.98
Packaging Poly Bottle
Chemical Name or Material Sodium hypophosphite hydrate
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'