missing translation for 'onlineSavingsMsg'
Learn More

Sodium Molybdate, ≥98%, Honeywell Fluka™

Product Code. p-7120812 Shop All Honeywell Fluka Products
Click to view available options
Quantity:
5 g
500 g
Packaging:
Poly Bottle
Unit Size:
500g
5g
This item is not returnable. View return policy

Product Code. 15622630

Brand: Honeywell-Fluka 2436555G

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

≥98%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7631-95-0
Molecular Formula MoNa2O4
Molecular Weight (g/mol) 205.926
MDL Number MFCD00003486
InChI Key TVXXNOYZHKPKGW-UHFFFAOYSA-N
Synonym sodium molybdate, disodium molybdate, sodium molybdate vi, natriummolybdat, molybdic acid, disodium salt, natriummolybdat german, sodium molybdate van, sodium molybdate na2moo4, unii-948qaq08i1, ccris 5442
PubChem CID 61424
ChEBI CHEBI:75215
IUPAC Name disodium;dioxido(dioxo)molybdenum
SMILES [O-][Mo](=O)(=O)[O-].[Na+].[Na+]

Specifications

Melting Point 687°C
Color White
Quantity 5 g
Linear Formula Na2MoO4
UN Number NONH for all modes of transport
Formula Weight 205.92g/mol
Percent Purity ≥98%
Packaging Poly Bottle
Appearance Granular powder
Density 3.78g/mL at 25°C
Chemical Name or Material Sodium Molybdate
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.