missing translation for 'onlineSavingsMsg'
Learn More

Sodium molybdenum oxide, anhydrous, Mo 46.2%

Product Code. 11336618
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
This item is not returnable. View return policy

Product Code. 11336618

Brand: Thermo Scientific Alfa Aesar 012209.36

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7631-95-0 | MoNa2O4 | 205.926 g/mol

It is used as an analytical reagent, corrosion inhibitor, and non-oxidizing anodic inhibitor. It reduces the nitrite requirement of fluids inhibited with nitrite-amine and improves the corrosion protection of carboxylate salt fluids. It is utilized in pigments, dyes, anti-freezing agent, cutting oils, metal finishing agent, brightening agent for zinc, additive for fertilizers and micronutrients.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7631-95-0
Molecular Formula MoNa2O4
Molecular Weight (g/mol) 205.926
MDL Number MFCD00003486
InChI Key TVXXNOYZHKPKGW-UHFFFAOYSA-N
Synonym sodium molybdate, disodium molybdate, sodium molybdate vi, natriummolybdat, molybdic acid, disodium salt, natriummolybdat german, sodium molybdate van, sodium molybdate na2moo4, unii-948qaq08i1, ccris 5442
PubChem CID 61424
ChEBI CHEBI:75215
IUPAC Name disodium;dioxido(dioxo)molybdenum
SMILES [O-][Mo](=O)(=O)[O-].[Na+].[Na+]

Specifications

Melting Point 687°C
Color White
Physical Form Powder
Quantity 500 g
Linear Formula Na2MoO4
Merck Index 14,8645
Solubility Information It is soluble in water.
Formula Weight 205.92
Concentration or Composition (by Analyte or Components) Mo 46.2%
Odor Odorless
Refractive Index 1.714
Sensitivity Hygroscopic
Density 3.72 g/mL
Chemical Name or Material Sodium molybdenum oxide, Anhydrous
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.