Learn More
Melford Sodium Phosphate Dibasic Heptahydrate
Description
Sodium Phosphate Dibasic Heptahydrate, also known as disodium hydrogen phosphate heptahydrate, is a chemical compound with significant applications in the life science marketplace.
It is a hydrated form of sodium phosphate dibasic, meaning it contains seven water molecules per molecule of the compound.
Sodium Phosphate Dibasic Heptahydrate is primarily used as a buffering agent and as a source of phosphate ions. Buffers help maintain a stable pH in biological and biochemical processes, ensuring optimal conditions for enzymatic reactions, cell culture, and other molecular biology techniques.
It is also commonly used in laboratory settings for the preparation of buffer solutions with specific pH ranges and buffering capacities. It is particularly useful in applications where a higher pH range is required, as it has a buffering capacity in the alkaline pH range
Moreover, the compound is used in various biological and biochemical assays, protein purification processes, and the formulation of diagnostic kits and pharmaceutical products. It serves as a source of phosphate ions, which are essential for numerous biological processes such as DNA and RNA synthesis, energy metabolism, and signal transduction.
Specifications
Specifications
| Chemical Name or Material | Sodium Phosphate Dibasic Heptahydrate |
| CAS | 7782-85-6 |
| Quantity | 500 g |
| Molecular Formula | Na2HPO4-7H2O |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.