missing translation for 'onlineSavingsMsg'
Learn More

Sodium Phosphate Dibasic Heptahydrate (Colorless-to-White Crystals), Fisher BioReagents™

Catalog Number p-28670
Click to view available options
Quantity:
500 g
1 kg
Packaging:
Glass Bottle
Poly Bottle
This item is not returnable. View return policy
7782-85-6
H17Na2O11P
270.08
MFCD00149180
PYLIXCKOHOHGKQ-UHFFFAOYSA-N
sodium phosphate dibasic heptahydrate, unii-70wt22sf4b, sodium phosphate, dibasic, heptahydrate, sodium hydrogen phosphate heptahydrate, disodium hydrogen phosphate heptahydrate, axea paragraph signthornaae, 2na.hpo4.7h2o, sodium hydrogenphosphate heptahydrate, sodium phosphate dibasic, acs, disodium hydrogenphosphate heptahydrate
6096963
disodium;hydrogen phosphate;heptahydrate
O.O.O.O.O.O.O.[Na+].[Na+].OP(O)(O)=O
This item is not returnable. View return policy
7782-85-6
H17Na2O11P
270.08
MFCD00149180
PYLIXCKOHOHGKQ-UHFFFAOYSA-N
sodium phosphate dibasic heptahydrate, unii-70wt22sf4b, sodium phosphate, dibasic, heptahydrate, sodium hydrogen phosphate heptahydrate, disodium hydrogen phosphate heptahydrate, axea paragraph signthornaae, 2na.hpo4.7h2o, sodium hydrogenphosphate heptahydrate, sodium phosphate dibasic, acs, disodium hydrogenphosphate heptahydrate
6096963
disodium;hydrogen phosphate;heptahydrate
O.O.O.O.O.O.O.[Na+].[Na+].OP(O)(O)=O

Sodium Phosphate Dibasic Heptahydrate (Colorless-to-White Crystals)

This buffer component is commonly used in tissue culture and molecular biology applications.
TRUSTED_SUSTAINABILITY
Melting Point 48°C
Color White
pH 9.0 to 9.3
Physical Form Solid
Quantity 500 g
Assay Percent Range 98.0 to 102.0 %
Merck Index 15, 8789
Formula Weight 268.07
Percent Purity 98.0 to 102.0%
Identification Pass Test
Packaging Glass Bottle
Assay 99.0-102.0%
Chemical Name or Material Sodium Phosphate Dibasic Heptahydrate
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'