missing translation for 'onlineSavingsMsg'
Learn More

Sodium phosphate monobasic monohydrate, puriss. p.a., ACS Reagent, Honeywell Fluka™

Catalog Number p-7072258
Click to view available options
Quantity:
250 g
1 kg
10049-21-5
H5NaO5P
139.00
MFCD00149208
BBMHARZCALWXSL-UHFFFAOYSA-N
sodium dihydrogen phosphate monohydrate, sodium phosphate monobasic monohydrate, monosodium phosphate monohydrate, unii-593yog76rn, phosphoric acid, monosodium salt, monohydrate, sodium phosphate monobasic hydrate, sodium dihydrogen phosphate hydrate, sodium dihydrogenphosphate monohydrate, sodium hydrate dihydrogen phosphate, pubchem12707
516949
sodium;dihydrogen phosphate;hydrate
O.[Na+].OP(O)(O)=O
10049-21-5
H5NaO5P
139.00
MFCD00149208
BBMHARZCALWXSL-UHFFFAOYSA-N
sodium dihydrogen phosphate monohydrate, sodium phosphate monobasic monohydrate, monosodium phosphate monohydrate, unii-593yog76rn, phosphoric acid, monosodium salt, monohydrate, sodium phosphate monobasic hydrate, sodium dihydrogen phosphate hydrate, sodium dihydrogenphosphate monohydrate, sodium hydrate dihydrogen phosphate, pubchem12707
516949
sodium;dihydrogen phosphate;hydrate
O.[Na+].OP(O)(O)=O

Puriss. p.a., ACS Reagent,≥99.0% (T)

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY
Quantity 250 g
Linear Formula NaH2PO4 · H2O
UN Number NONH for all modes of transport
Formula Weight 137.99g/mol
Percent Purity ≥99% (T)
Grade ACS/puriss. p.a.
Chemical Name or Material Sodium phosphate monobasic monohydrate
Product Identifier
  • Sodium phosphate monobasic monohydrate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'