Learn More
Sodium pyrophosphate tetrabasic decahydrate, Puriss. p.a., ≥99%, Honeywell Fluka™
Puriss. p.a.,≥99%
Brand: Honeywell Fluka 30411-50KG
Additional Details : CAS : 13472-36-1 Poids : 50.00000kg
Quantity | 50kg |
---|
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Chemical Identifiers
13472-36-1 | |
446.05 | |
VZWGHDYJGOMEKT-UHFFFAOYSA-J | |
3084150 | |
O.O.O.O.O.O.O.O.O.O.[O-]P(=O)([O-])OP(=O)([O-])[O-].[Na+].[Na+].[Na+].[Na+] |
H20Na4O17P2 | |
MFCD00003524 | |
sodium pyrophosphate decahydrate, unii-iy3dkb96qw, tetrasodium pyrophosphate decahydrate, diphosphoric acid, tetrasodium salt, decahydrate, iy3dkb96qw, pyrophosphate sodium salt decahydrate, pyrophosphate inorganic, colostrum, sodium phosphate pyro, sodium pyrophosphate na4p2o7 decahydrate | |
tetrasodium;phosphonato phosphate;decahydrate |
Specifications
Sodium pyrophosphate tetrabasic decahydrate | |
13472-36-1 | |
H20Na4O17P2 | |
MFCD00003524 | |
sodium pyrophosphate decahydrate, unii-iy3dkb96qw, tetrasodium pyrophosphate decahydrate, diphosphoric acid, tetrasodium salt, decahydrate, iy3dkb96qw, pyrophosphate sodium salt decahydrate, pyrophosphate inorganic, colostrum, sodium phosphate pyro, sodium pyrophosphate na4p2o7 decahydrate | |
O.O.O.O.O.O.O.O.O.O.[O-]P(=O)([O-])OP(=O)([O-])[O-].[Na+].[Na+].[Na+].[Na+] | |
446.05 | |
446.06 | |
puriss. p.a. | |
Fiberboard Box |
Puriss. p.a., ≥99% | |
100.0000% | |
NaF | |
NONH for all modes of transport | |
VZWGHDYJGOMEKT-UHFFFAOYSA-J | |
tetrasodium;phosphonato phosphate;decahydrate | |
3084150 | |
>99% | |
1.820 g/cm3 | |
50kg |