missing translation for 'onlineSavingsMsg'
Learn More

Sodium pyrophosphate tetrabasic decahydrate, Puriss. p.a., ≥99%, Honeywell Fluka™

Puriss. p.a.,≥99%

Brand:  Honeywell Fluka 30411-2.5KG

Additional Details : CAS : 13472-36-1 Poids : 2.50000kg

Quantity 2.5kg

 View more versions of this product

Product Code. 15643020

  • 134.00€ / 2.50kg


En stock, prêt à être expédié : 20
Add to basket

Description

Description

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications

Chemical Identifiers

13472-36-1
446.05
VZWGHDYJGOMEKT-UHFFFAOYSA-J
3084150
O.O.O.O.O.O.O.O.O.O.[O-]P(=O)([O-])OP(=O)([O-])[O-].[Na+].[Na+].[Na+].[Na+]
H20Na4O17P2
MFCD00149200
sodium pyrophosphate decahydrate, unii-iy3dkb96qw, tetrasodium pyrophosphate decahydrate, diphosphoric acid, tetrasodium salt, decahydrate, iy3dkb96qw, pyrophosphate sodium salt decahydrate, pyrophosphate inorganic, colostrum, sodium phosphate pyro, sodium pyrophosphate na4p2o7 decahydrate
tetrasodium;phosphonato phosphate;decahydrate

Specifications

Sodium Pyrophosphate Tetrabasic Decahydrate
H20Na4O17P2
MFCD00149200
sodium pyrophosphate decahydrate, unii-iy3dkb96qw, tetrasodium pyrophosphate decahydrate, diphosphoric acid, tetrasodium salt, decahydrate, iy3dkb96qw, pyrophosphate sodium salt decahydrate, pyrophosphate inorganic, colostrum, sodium phosphate pyro, sodium pyrophosphate na4p2o7 decahydrate
O.O.O.O.O.O.O.O.O.O.[O-]P(=O)([O-])OP(=O)([O-])[O-].[Na+].[Na+].[Na+].[Na+]
446.05
446.06g/mol
puriss. p.a.
13472-36-1
Na4P2O7 · 10H2O
NONH for all modes of transport
VZWGHDYJGOMEKT-UHFFFAOYSA-J
tetrasodium;phosphonato phosphate;decahydrate
3084150
≥99%
2.5kg
Videos
SDS Documents