missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Selenite, ∽98%, ('White to off-white powder), MP Biomedicals
Shop All MP Biomedicals ProductsSpecifications
Specifications
| Color | White |
| Physical Form | Powder |
| Quantity | 100 g |
| Assay Percent Range | ∽98% |
| Formula Weight | 172.9 |
| Chemical Name or Material | Sodium Selenite |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?