missing translation for 'onlineSavingsMsg'
Learn More

Sodium Solution for ICP, 1g/L H2O Matrix, Fisher Chemical™
BIOPHARMA

Product Code. 12923454 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
100 mL
250 mL
500 mL
Unit Size:
100mL
250mL
500mL
Product Code. Quantity unitSize
12923454 100 mL 100mL
12933454 250 mL 250mL
12943454 500 mL 500mL
3 options
This item is not returnable. View return policy

Product Code. 12923454

Brand: Fisher Chemical S 035 100

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.

Specifications

Solution Contains H2O
Concentration 1 g/L
Quantity 100 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.