missing translation for 'onlineSavingsMsg'
Learn More

Sodium solution for ICP/MS 10mg/L HNO3 matrix, Fisher Chemical™

Catalog Number 12967383
Click to view available options
Quantity:
100 mL
Packaging:
Bottle
This item is not returnable. View return policy
7440-23-5
Na
22.99
MFCD00085307
KEAYESYHFKHZAL-UHFFFAOYSA-N
5360545
CHEBI:26708
sodium
[Na]
This item is not returnable. View return policy
7440-23-5
Na
22.99
MFCD00085307
KEAYESYHFKHZAL-UHFFFAOYSA-N
5360545
CHEBI:26708
sodium
[Na]

All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.

Concentration 10mg/L
Packaging Bottle
Quantity 100 mL
Solvent or Matrix HNO3
Chemical Name or Material Sodium
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'