missing translation for 'onlineSavingsMsg'
Learn More

Sodium Standard Metal Solution 1000ppm, for AAS, Solution HNO3 1M, Fisher Chemical™

Catalog Number p-7073222
Click to view available options
Quantity:
100 mL
500 mL
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
85307
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
85307

Sodium Standard Metal Solution 1000ppm, for AAS, Solution HNO3 1M

TRUSTED_SUSTAINABILITY
Color Colorless
pH 1
Quantity 100 mL
Packaging HDPE plastic bottle
Form Liquid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'