Learn More
Honeywell Fluka™ Sodium Sulfate, ACS Reagent, Anhydrous Powder, Honeywell Fluka™
ACS Reagent, anhydrous, powder,≥99.0%
Brand: Honeywell Fluka™ 238597-25G
Additional Details : CAS : 7757-82-6 Poids : 0.02500kg
Quantity | 25g |
---|
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Chemical Identifiers
7757-82-6 | |
142.036 | |
PMZURENOXWZQFD-UHFFFAOYSA-L | |
24436 | |
disodium;sulfate |
Na2O4S | |
MFCD00003504 | |
sodium sulfate, disodium sulfate, salt cake, thenardite, sodium sulfate anhydrous, sodium sulphate, sodium sulfate, anhydrous, sulfuric acid disodium salt, disodium sulphate, sodium sulfate 2:1 | |
CHEBI:32149 | |
[O-]S(=O)(=O)[O-].[Na+].[Na+] |
Specifications
Sodium Sulfate | |
Na2O4S | |
MFCD00003504 | |
sodium sulfate, disodium sulfate, salt cake, thenardite, sodium sulfate anhydrous, sodium sulphate, sodium sulfate, anhydrous, sulfuric acid disodium salt, disodium sulphate, sodium sulfate 2:1 | |
[O-]S(=O)(=O)[O-].[Na+].[Na+] | |
142.036 | |
CHEBI:32149 | |
≥99% | |
2.68g/mL at 25°C | |
25g |
7757-82-6 | |
Na2SO4 | |
NONH for all modes of transport | |
PMZURENOXWZQFD-UHFFFAOYSA-L | |
disodium;sulfate | |
24436 | |
142.04g/mol | |
ACS Reagent | |
Anhydrous Powder |