missing translation for 'onlineSavingsMsg'
Learn More

Sodium Sulfate, ACS Reagent, Anhydrous Powder, Honeywell Fluka™

Catalog Number p-7071924
Click to view available options
Quantity:
25 g
500 g
1 kg
2.5 kg
5 kg
12 kg
This item is not returnable. View return policy
7757-82-6
Na2O4S
142.04
MFCD00003504
PMZURENOXWZQFD-UHFFFAOYSA-L
sodium sulfate, disodium sulfate, salt cake, thenardite, sodium sulfate anhydrous, sodium sulphate, sodium sulfate, anhydrous, sulfuric acid disodium salt, disodium sulphate, sodium sulfate 2:1
24436
CHEBI:32149
disodium;sulfate
[Na+].[Na+].[O-]S([O-])(=O)=O
This item is not returnable. View return policy
7757-82-6
Na2O4S
142.04
MFCD00003504
PMZURENOXWZQFD-UHFFFAOYSA-L
sodium sulfate, disodium sulfate, salt cake, thenardite, sodium sulfate anhydrous, sodium sulphate, sodium sulfate, anhydrous, sulfuric acid disodium salt, disodium sulphate, sodium sulfate 2:1
24436
CHEBI:32149
disodium;sulfate
[Na+].[Na+].[O-]S([O-])(=O)=O

ACS Reagent, anhydrous, powder,≥99.0%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY
Physical Form Anhydrous Powder
Quantity 25 g
Linear Formula Na2SO4
UN Number NONH for all modes of transport
Formula Weight 142.04g/mol
Percent Purity ≥99%
Grade ACS Reagent
Density 2.68g/mL at 25°C
Chemical Name or Material Sodium Sulfate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'