missing translation for 'onlineSavingsMsg'
Learn More

Melford Sodium Sulfate, Anhydrous

Product Code. 30018809
Click to view available options
Quantity:
500 g
1 kg
5 kg
Unit Size:
1kg
500g
5kg
This item is not returnable. View return policy

Product Code. 30018809

Brand: Melford S25150500.0

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Fine white granular form.

Fine white granular form. Store at room temperature.

Specifications

Chemical Name or Material Sodium Sulfate, Anhydrous
CAS 7757-82-6
Quantity 500 g
Molecular Formula Na2SO4
Grade ACS
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.