missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium sulfate, Free-flowing, Reagent Grade, anhydrous, ≥99%, Honeywell Fluka™
Click to view available options
Quantity:
500 g
1 kg
2.5 kg
5 kg
12 kg
Packaging:
Fiberboard Box
Poly Bottle
Description
- ≥95% for general laboratory use.
Specifications
Melting Point | 884°C (literature) |
CAS Max % | 100.0000% |
Quantity | 500 g |
Linear Formula | Na2SO4 |
Formula Weight | 142.04 |
Percent Purity | >99% |
Grade | Reagent |
Packaging | Poly Bottle |
Density | 2.68 g/mL at 25 °C(lit.) |
Chemical Name or Material | Sodium sulfate, Free-flowing, Reagent Grade, anhydrous, ≥99% |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'