Learn More
Honeywell Fluka™ Sodium sulfate, Free-flowing, Reagent Grade, anhydrous, ≥99%, Honeywell Fluka™
Free-flowing, Reagent Grade, anhydrous,≥99%
Brand: Honeywell Fluka™ 793531-1KG
Additional Details : CAS : 7757-82-6 Poids : 1.00000kg
Packaging | Poly Bottle |
---|---|
Quantity | 1kg |
Description
- ≥95% for general laboratory use.
Chemical Identifiers
7757-82-6 | |
142.036 | |
PMZURENOXWZQFD-UHFFFAOYSA-L | |
24436 | |
disodium;sulfate |
Na2O4S | |
MFCD00003504 | |
sodium sulfate, disodium sulfate, salt cake, thenardite, sodium sulfate anhydrous, sodium sulphate, sodium sulfate, anhydrous, sulfuric acid disodium salt, disodium sulphate, sodium sulfate 2:1 | |
CHEBI:32149 | |
[O-]S(=O)(=O)[O-].[Na+].[Na+] |
Specifications
Sodium sulfate | |
7757-82-6 | |
Na2O4S | |
MFCD00003504 | |
PMZURENOXWZQFD-UHFFFAOYSA-L | |
disodium;sulfate | |
24436 | |
142.04 | |
Reagent | |
Poly Bottle | |
1kg |
Free-flowing, Reagent Grade, anhydrous, ≥99% | |
100.0000% | |
Na2SO4 | |
sodium sulfate, disodium sulfate, salt cake, thenardite, sodium sulfate anhydrous, sodium sulphate, sodium sulfate, anhydrous, sulfuric acid disodium salt, disodium sulphate, sodium sulfate 2:1 | |
[O-]S(=O)(=O)[O-].[Na+].[Na+] | |
142.036 | |
CHEBI:32149 | |
>99.0% | |
2.68 g/mL at 25 °C(lit.) | |
884°C (literature) |