missing translation for 'onlineSavingsMsg'
Learn More

Sodium sulfite, Honeywell Fluka™

Catalog Number p-7072309
Click to view available options
Quantity:
250 g
2.5 kg
This item is not returnable. View return policy
7757-83-7
Na2O3S
126.037
MFCD00003503
GEHJYWRUCIMESM-UHFFFAOYSA-L
sodium sulfite, disodium sulfite, sodium sulphite, sodium sulfite anhydrous, sulfurous acid, disodium salt, sulftech, natrium sulfurosum, sodiumsulfite, natriumsulfid, natriumsulfit
24437
CHEBI:86477
disodium;sulfite
[O-]S(=O)[O-].[Na+].[Na+]
This item is not returnable. View return policy
7757-83-7
Na2O3S
126.037
MFCD00003503
GEHJYWRUCIMESM-UHFFFAOYSA-L
sodium sulfite, disodium sulfite, sodium sulphite, sodium sulfite anhydrous, sulfurous acid, disodium salt, sulftech, natrium sulfurosum, sodiumsulfite, natriumsulfid, natriumsulfit
24437
CHEBI:86477
disodium;sulfite
[O-]S(=O)[O-].[Na+].[Na+]

≥98%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY
Quantity 250 g
Linear Formula Na2SO3
UN Number NONH for all modes of transport
Formula Weight 126.04g/mol
Percent Purity ≥98%
Chemical Name or Material Sodium sulfite
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'