missing translation for 'onlineSavingsMsg'
Learn More

Sodium Tetraborate Decahydrate, puriss. p.a. ACS Reagent, Honeywell Fluka™

Catalog Number p-7072034
Click to view available options
Quantity:
100 g
500 g
1 kg
5 kg
25 kg
This item is not returnable. View return policy
1303-96-4
MFCD00149193
This item is not returnable. View return policy
1303-96-4
MFCD00149193

Puriss. p.a., ACS Reagent, Reagent ISO, buffer substance,≥99.5%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY
Quantity 100 g
Linear Formula Na2B4O7 · 10H2O
UN Number NONH for all modes of transport
Formula Weight 381.37g/mol
Percent Purity ≥99.5%
Grade ACS/puriss. p.a.
Density 1.73g/mL at 25°C
Chemical Name or Material Sodium Tetraborate Decahydrate, Reag. ISO
compliance-icons
Product Identifier
  • Sodium tetraborate decahydrate
Signal Word
  • Danger
Hazard Category
  • Reproductive toxicity Category 1B
Hazard Statement
  • H360FD-May damage fertility. May damage the unborn child.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'