missing translation for 'onlineSavingsMsg'
Learn More

Sodium Thiocyanate, 98 to 102% (titration), Reagent Grade, Honeywell™

Product Code. p-7120837 Shop All Honeywell Research Chemicals Products
Click to view available options
Quantity:
250 g
1 kg
3 kg
Packaging:
Poly bottle
Unit Size:
1kg
250g
3kg
This item is not returnable. View return policy

Product Code. 15648110

Brand: Honeywell Research Chemicals S7757250G

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

Reagent Grade, 98-102% (titration)

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 540-72-7
Molecular Formula CNNaS
Molecular Weight (g/mol) 81.07
MDL Number MFCD00011123
InChI Key VGTPCRGMBIAPIM-UHFFFAOYSA-M
Synonym sodium thiocyanate, sodium rhodanate, sodium sulfocyanate, sodium rhodanide, thiocyanic acid, sodium salt, thiocyanate sodium, scyan, sodium isothiocyanate, nascn, haimased
PubChem CID 516871
ChEBI CHEBI:30952
SMILES [Na+].[S-]C#N

Specifications

Melting Point 308°C
Color Colorless
Physical Form Solid
Quantity 250 g
Linear Formula NaSCN
UN Number NONH for all modes of transport
Beilstein 3594965
IUPAC Name sodium;thiocyanate
Formula Weight 81.07g/mol
Percent Purity 98 to 102% (titration)
Odor Odorless
Packaging Poly bottle
Density 1,76 g/cu.cm at 20°C
Chemical Name or Material Sodium thiocyanate
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.