missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Notrax Sof-Tred™ Microcellular Vinyl Foam with Pebble Pattern, Blue
Click to view available options
Unit Size:
Each
Description
- Pebble pattern provides non-directional traction and is easy to clean
- Basic worker comfort derived from 94 mm thick worker platform comprised of a construction of microcellular vinyl consisting of millions of air bubbles that provide basic ergonomic benefit
- All four sides are bevelled and sealed to minimize tripping hazards
Specifications
Specifications
| Color | Blue |
| Length (English) | 36 in. |
| Length (Metric) | 91 cm |
| Material | Microcellular Vinyl |
| Type | Industrial and Commercial Matting |
| Width (Metric) | 60 cm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction