Learn More
Sampling systems Soil Auger Sampler - 1000mm
Brand:  Sampling systems 1624A-1050
Soil Auger Sampler - 1000mm
Product Code. 18815443
  • ONLINE EXCLUSIVE PRICE
    421.58€
    301.13€ / Each
    Save 120.45€
    28% Off