missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Solvent Blue 38

Product Code. p-7022559
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 11418356

Brand: Thermo Scientific Chemicals A15395.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1328-51-4 | C32H14CuN8Na2O6S2 | 780.16 g/mol

Solvent Blue 38 is commonly utilized in microscopy to detect demyelination in the central nervous system. It is used as a myelin-sheath stain.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Solvent Blue 38 is commonly utilized in microscopy to detect demyelination in the central nervous system. It is used as a myelin-sheath stain.

Solubility
Soluble in water (partly), and methanol (1 mg/ml).

Notes
Store in cool, dry conditions in a well sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1328-51-4
Molecular Formula C32H14CuN8Na2O6S2
Molecular Weight (g/mol) 780.16
MDL Number MFCD00071424
InChI Key DKBXPLYSDKSFEQ-UHFFFAOYSA-L
PubChem CID 92030797
SMILES C1=CC=C2C(=C1)C3=NC4=C5C=CC=CC5=C([N-]4)N=C6C7=C(C=CC=C7S(=O)(=O)[O-])C(=N6)N=C8C9=C(C(=CC=C9)S(=O)(=O)[O-])C(=N8)N=C2[N-]3.[Na+].[Na+].[Cu+2]

Specifications

Melting Point ∼235°C (decomposition)
Solubility Information Soluble in water (partly),and methanol (1mg/ml).
Quantity 25 g
Chemical Name or Material Fast Blue

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.