Learn More
Horiba™ S-316 Solvent Extractant
Click to view available options
Quantity:
1.5 kg
Unit Size:
Each
Description
- 1.5kg bottle
- Does not contain carbon tetrachloride
- Quantitative measurement by infrared spectrophotometry
Specifications
Specifications
| For Use With (Equipment) | SR-305 Oil Content Analyzer |
| Packaging | 1.5 kg Bottle |
| Quantity | 1.5 kg |
| Type | Solvent |