Learn More
Qsonica Q800R3 Sonicator
Product Code. 15986095 Shop All Fisherbrand Products
Click to view available options
Voltage:
110 V
Unit Size:
Each
Regulatory: This product is under eco-contribution tax.
Product Code. 15986095
Only null left
Add to basket
Add to basket
Regulatory: This product is under eco-contribution tax.
DNA Shearing Sonicator
Q800R3 is a complete package including a chiller to control temperature, digital operating system with programmable memory, high intensity ultrasonics and sound reducing enclosure.
  • Samples can be sheared to a range of fragment sizes (150bp – 3kb)
  • System is compatible with commercially available sample tubes and is capable of processing up to 18 samples at one time
  • Multiple tube rack options can accommodate 50μL to 1ml sample volumes using a variety of standard sample tubes
Specifications
Product Type DNA and Chromatin Shearing System
Voltage 110 V
Hertz 20 kHz
Wattage 750 W
Dimensions (L x W x H) 15.25 x 8 x 8.5 in.
For Use With (Application) DNA shearing for Next-gen sequencing, Chromatin shearing, ChIP, ChIP-seq, RNA-seq, Protein extraction, Cell lysis
Includes Generator, Converter, Ultrasonic Horn, Rotation System, Chiller, Cables, Tubing, 12 x 0.5mL tube rack, 2-year warranty
Timer 1 sec. to 10 hrs.