Learn More
SP Bel-Art™ SP Bel-Art WYE (Y) Tubing Connecters For 5/16 in. Tubing
Use these fittings to ensure a tight, secure connection between lengths of tubing having the same inner diameter
Brand: SP Bel-Art™ 19615-0000
Additional Details : Weight : 0.24300kg
Description
- Multiple barbs per arm provide additional contact between the fitting and the tubing for extra gripping power
- Chemical-resistant polypropylene plastic for use with a wide range of liquids or gases
- FDA-grade for direct contact with food, drugs, and cosmetics
- Autoclavable
Specifications
8 mm | |
39.7 mm | |
54 mm |
Polypropylene | |
12/Pk. |