Learn More
BRAND™ BlauBRAND™ Burette Tube for Compact Burette
Borosilicate glass construction. Replacement burette tube for compact burette.
Brand: BRAND™ 13943
32.62 EUR valid until 2024-12-31
Use promo code "23264" to get your promotional price.
Additional Details : Weight : 0.05000kg
Description
- BLAUBRAND™ borosilicate glass (3.3) construction
- Replacement burette tube
Specifications
10 mL | |
Borosilicate Glass | |
Buret Tube |
Compact Burette | |
1 Each |