Learn More
Ebro Electronic™ SSX 210 Salt Meter Probe
Product Code. 11793569
Click to view available options
Display Type:
No Display
Unit Size:
Each
Product Code. 11793569
Brand: Ebro Electronic™ 13415210
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Exchange probe for SSX 210, Probe SSX 210 with 150cm cable
Used to measure the salt content in fluid and semi-solid food products, such as meat, cold cuts, cheese, salads etc.
Order Info
Type
TRUSTED_SUSTAINABILITY
Specifications
Type Probe
Display Type No Display
Calibration Not Calibrated
Alarm Types None
Model SPX 210