Learn More
SI Analytics™ Spear Tip pH Combination Electrode
Guarentees high measurement accuracy and stability as well as long service life. SI Analytics™ Spear Tip pH Combination Electrode is highly adaptable to measuring tasks.
287.00€ - 330.00€
Specifications
Material | A-Glass |
---|---|
Type | pH Combination Electrode |
Internal Reference | Silamid™ |
Product Code | Brand | Junction | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Junction | Price | Quantity & Availability | |||||
10305441
|
SI Analytics™
285101211 |
3 x Ceramic Diaphragm |
330.00€
Each |
Estimated Shipment: 04-07-2024
Log in to see stock. |
|||||
10335581
|
SI Analytics™
285101285 |
Hole Diaphragm |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance. |
||||||
Description
- Plug head connection
- Spear tip
- Constructed of A-Glass
Specifications
A-Glass | |
pH Combination Electrode | |
Silamid™ |